Product Details

SNP ID
rs201626107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127507297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGCTGGGGCTGCCGCCCCCGC[C/T]GCTGTTGCTGTACTCCTCGCCCCAG
Phenotype
MIM: 614045 MIM: 610933
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM129B PubMed Links

Gene Details

Gene
FAM129B
Gene Name
family with sequence similarity 129 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035534.2 1820 Missense Mutation AGC,GGC S584G NP_001030611.1
NM_022833.3 1820 Missense Mutation AGC,GGC S597G NP_073744.2
XM_005252135.2 1820 Missense Mutation AGC,GGC S622G XP_005252192.2
XM_011518925.1 1820 Missense Mutation AGC,GGC S627G XP_011517227.1
Gene
LRSAM1
Gene Name
leucine rich repeat and sterile alpha motif containing 1
There are no transcripts associated with this gene.

View Full Product Details