Product Details

SNP ID
rs199934348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:20015282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATAATCTGTTCTCTCTCGAGAC[G/T]CATCTGTTCAGCTACCTCCCGGGCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAP7D2 PubMed Links

Gene Details

Gene
MAP7D2
Gene Name
MAP7 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168465.1 1579 Missense Mutation NP_001161937.1
NM_001168466.1 1579 Missense Mutation NP_001161938.1
NM_001168467.1 1579 Missense Mutation NP_001161939.1
NM_152780.3 1579 Missense Mutation NP_689993.2
XM_005274478.2 1579 Missense Mutation XP_005274535.1
XM_005274479.2 1579 Missense Mutation XP_005274536.1
XM_005274480.3 1579 Missense Mutation XP_005274537.1
XM_006724481.2 1579 Missense Mutation XP_006724544.1
XM_006724482.2 1579 Missense Mutation XP_006724545.1
XM_011545483.1 1579 Missense Mutation XP_011543785.1
XM_011545484.1 1579 Missense Mutation XP_011543786.1
XM_011545485.1 1579 Missense Mutation XP_011543787.1
XM_011545486.2 1579 Intron XP_011543788.1
XM_017029393.1 1579 Missense Mutation XP_016884882.1
XM_017029394.1 1579 Missense Mutation XP_016884883.1
XM_017029395.1 1579 Missense Mutation XP_016884884.1
XM_017029396.1 1579 Missense Mutation XP_016884885.1
XM_017029397.1 1579 Missense Mutation XP_016884886.1
XM_017029398.1 1579 Intron XP_016884887.1
Gene
MIR23C
Gene Name
microRNA 23c
There are no transcripts associated with this gene.

View Full Product Details