Product Details

SNP ID
rs199726768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47142419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTATTTCACAAGCCTCTCAGCTTC[A/G]CGGCGGGACCACTCTTTCATCCTGG
Phenotype
MIM: 300403 MIM: 300080
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDUFB11 PubMed Links

Gene Details

Gene
NDUFB11
Gene Name
NADH:ubiquinone oxidoreductase subunit B11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135998.2 891 Silent Mutation CGC,CGT R120R NP_001129470.1
NM_019056.6 891 Silent Mutation CGC,CGT R130R NP_061929.2
Gene
RBM10
Gene Name
RNA binding motif protein 10
There are no transcripts associated with this gene.

View Full Product Details