Product Details

SNP ID
rs199858408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103825384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCAGGTTTGTGCCTCCTCAGTA[C/G]TCACTTGCCTATCCTCTTTGTCTAC
Phenotype
MIM: 300285
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RAB9B PubMed Links

Gene Details

Gene
RAB9B
Gene Name
RAB9B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016370.2 740 Missense Mutation ACT,AGT T134S NP_057454.1

View Full Product Details