Product Details

SNP ID
rs200360009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106205522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTGAAACTTCATCAAACAGTAA[G/T]AGGAAAAAGGCATTTTCTCTAGAAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MUM1L1 PubMed Links

Gene Details

Gene
MUM1L1
Gene Name
MUM1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171020.1 733 Missense Mutation AAG,AAT K30N NP_001164491.1
NM_152423.4 733 Missense Mutation AAG,AAT K30N NP_689636.3
XM_005262077.2 733 Missense Mutation AAG,AAT K30N XP_005262134.1
XM_011530856.1 733 Missense Mutation AAG,AAT K30N XP_011529158.1
XM_011530857.1 733 Missense Mutation AAG,AAT K30N XP_011529159.1
XM_017029269.1 733 Missense Mutation AAG,AAT K30N XP_016884758.1

View Full Product Details