Product Details

SNP ID
rs199939747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:34130599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGATGGGACTCTGGAGCTTTGGG[A/G]GGCTCCGGGTGGAGACTGGACACCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM47A PubMed Links

Gene Details

Gene
FAM47A
Gene Name
family with sequence similarity 47 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203408.3 Intron NP_981953.2

View Full Product Details