Product Details

SNP ID
rs200018441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47241612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACTCTCAGGAGCTGCTGTCATT[C/T]CTCCTGGACGGGCTGCATGAGGACC
Phenotype
MIM: 300050
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP11 PubMed Links

Gene Details

Gene
USP11
Gene Name
ubiquitin specific peptidase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004651.3 1221 Silent Mutation TTC,TTT F407F NP_004642.2
XM_005272674.3 1221 Silent Mutation TTC,TTT F134F XP_005272731.1
XM_011543988.1 1221 Silent Mutation TTC,TTT F134F XP_011542290.1

View Full Product Details