Product Details

SNP ID
rs200047366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:34131299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTCTCCAGTCTCCGGAGGCTCC[C/G]GGCGGAGACTGGACACCGGAGTCTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM47A PubMed Links

Gene Details

Gene
FAM47A
Gene Name
family with sequence similarity 47 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203408.3 Intron NP_981953.2

View Full Product Details