Product Details

SNP ID
rs199605232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47638973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGGTGCTCCTGCTCCCCGAGGG[C/G]GGCGCTGCTGCCCCTGCAGGAGCTG
Phenotype
MIM: 300383 MIM: 311040
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CFP PubMed Links

Gene Details

Gene
CFP
Gene Name
complement factor properdin
There are no transcripts associated with this gene.

Gene
ELK1
Gene Name
ELK1, ETS transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114123.2 792 Silent Mutation CCC,CCG P192P NP_001107595.1
NM_001257168.1 792 Intron NP_001244097.1
NM_005229.4 792 Silent Mutation CCC,CCG P192P NP_005220.2
XM_017029339.1 792 Silent Mutation CCC,CCG P192P XP_016884828.1

View Full Product Details