Product Details

SNP ID
rs199736694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103216393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACAATGAAGCGAGAGATGATGT[A/G]GAAAGGTTTGTAGGGCAGATGATGG
Phenotype
MIM: 300692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BEX4 PubMed Links

Gene Details

Gene
BEX4
Gene Name
brain expressed X-linked 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080425.3 392 Silent Mutation GTA,GTG V80V NP_001073894.1
NM_001127688.2 392 Silent Mutation GTA,GTG V80V NP_001121160.1

View Full Product Details