Product Details

SNP ID
rs199974088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152135490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGAGGATGGAAAAGAGGAGCTG[A/G]TGGAAGTGGATGATGAAGCATCCTC
Phenotype
MIM: 300343
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGEA10 PubMed Links

Gene Details

Gene
MAGEA10
Gene Name
MAGE family member A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011543.2 564 Missense Mutation ACC,ATC T44I NP_001011543.2
NM_001251828.1 564 Missense Mutation ACC,ATC T44I NP_001238757.1
NM_021048.4 564 Missense Mutation ACC,ATC T44I NP_066386.2
Gene
MAGEA10-MAGEA5
Gene Name
MAGEA10-MAGEA5 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204811.2 564 Intron NP_001191740.1

View Full Product Details