Product Details
- SNP ID
-
rs200107563
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:112779416 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGAGCAGCAGCAGCAGCAACAGCA[A/G]CTGGAGCAGCAGCTACCATGGTGGT
- Phenotype
-
MIM: 300410
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AMOT
PubMed Links
Gene Details
- Gene
- AMOT
- Gene Name
- angiomotin
- Gene
- MIR4329
- Gene Name
- microRNA 4329
There are no transcripts associated with this gene.
View Full Product Details