Product Details
- SNP ID
-
rs200133821
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:73447492 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGGCTCACCCTACAGTCCCCCGC[A/G]AGAAGACTGGAGCGTGTATCCTGGG
- Phenotype
-
MIM: 300025
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDX4
PubMed Links
Gene Details
- Gene
- CDX4
- Gene Name
- caudal type homeobox 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005193.1 |
239 |
Missense Mutation |
CAA,CGA |
Q80R |
NP_005184.1 |
View Full Product Details