Product Details

SNP ID
rs199870160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:87518107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAGCAACAGTCCTGTCCACCAC[A/G]ATATACTGGCACCAGTGCCAGGACC
Phenotype
MIM: 300348
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL4 PubMed Links

Gene Details

Gene
KLHL4
Gene Name
kelch like family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019117.4 396 Missense Mutation AAT,GAT N72D NP_061990.2
NM_057162.2 396 Missense Mutation AAT,GAT N72D NP_476503.1

View Full Product Details