Product Details

SNP ID
rs200265489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15385030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTCCCAATCTTTGATTTCCAGG[A/T]TTTGGCCCTTTCAAACCCATTTTTT
Phenotype
MIM: 300091 MIM: 300931
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FIGF PubMed Links

Gene Details

Gene
FIGF
Gene Name
c-fos induced growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004469.4 1332 Intron NP_004460.1
Gene
PIR
Gene Name
pirin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018109.2 1332 Missense Mutation ACC,TCC T283S NP_001018119.1
NM_003662.3 1332 Missense Mutation ACC,TCC T283S NP_003653.1
Gene
PIR-FIGF
Gene Name
PIR-FIGF readthrough
There are no transcripts associated with this gene.

View Full Product Details