Product Details

SNP ID
rs199638285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:78273018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATATGATATGGCATGAAACTGAC[A/T]AAAAAGGCAGCGGTCACGACCATGA
Phenotype
MIM: 300201
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYSLTR1 PubMed Links

Gene Details

Gene
CYSLTR1
Gene Name
cysteinyl leukotriene receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282186.1 1143 Missense Mutation TTA,TTT L243F NP_001269115.1
NM_001282187.1 1143 Missense Mutation TTA,TTT L243F NP_001269116.1
NM_001282188.1 1143 Missense Mutation TTA,TTT L243F NP_001269117.1
NM_006639.3 1143 Missense Mutation TTA,TTT L243F NP_006630.1

View Full Product Details