Product Details

SNP ID
rs200058043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:74304398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAATGTGGTTCCACCACCCTATC[C/T]TACACCTGTTACTGCTGTGGTGAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZCCHC13 PubMed Links

Gene Details

Gene
ZCCHC13
Gene Name
zinc finger CCHC-type containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203303.2 209 Silent Mutation TCC,TCT S44S NP_976048.1

View Full Product Details