Product Details
- SNP ID
-
rs200332017
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:47448076 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAGATTTCTGATGTATTTTGAGG[C/T]GCGACTTCCATATGAAGGCTTTTCC
- Phenotype
-
MIM: 314995
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF41
PubMed Links
Gene Details
- Gene
- ZNF41
- Gene Name
- zinc finger protein 41
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001324139.1 |
2461 |
Missense Mutation |
CAC,CGC |
H479R |
NP_001311068.1 |
NM_001324140.1 |
2461 |
Missense Mutation |
CAC,CGC |
H565R |
NP_001311069.1 |
NM_001324141.1 |
2461 |
Missense Mutation |
CAC,CGC |
H479R |
NP_001311070.1 |
NM_001324142.1 |
2461 |
Missense Mutation |
CAC,CGC |
H567R |
NP_001311071.1 |
NM_001324143.1 |
2461 |
Missense Mutation |
CAC,CGC |
H479R |
NP_001311072.1 |
NM_001324144.1 |
2461 |
Missense Mutation |
CAC,CGC |
H565R |
NP_001311073.1 |
NM_001324145.1 |
2461 |
Missense Mutation |
CAC,CGC |
H479R |
NP_001311074.1 |
NM_001324147.1 |
2461 |
Missense Mutation |
CAC,CGC |
H565R |
NP_001311076.1 |
NM_001324148.1 |
2461 |
Missense Mutation |
CAC,CGC |
H567R |
NP_001311077.1 |
NM_001324149.1 |
2461 |
Missense Mutation |
CAC,CGC |
H479R |
NP_001311078.1 |
NM_001324150.1 |
2461 |
Missense Mutation |
CAC,CGC |
H565R |
NP_001311079.1 |
NM_001324151.1 |
2461 |
Missense Mutation |
CAC,CGC |
H575R |
NP_001311080.1 |
NM_001324152.1 |
2461 |
Missense Mutation |
CAC,CGC |
H479R |
NP_001311081.1 |
NM_001324153.1 |
2461 |
Missense Mutation |
CAC,CGC |
H575R |
NP_001311082.1 |
NM_001324154.1 |
2461 |
Missense Mutation |
CAC,CGC |
H599R |
NP_001311083.1 |
NM_001324155.1 |
2461 |
Missense Mutation |
CAC,CGC |
H607R |
NP_001311084.1 |
NM_001324156.1 |
2461 |
Missense Mutation |
CAC,CGC |
H531R |
NP_001311085.1 |
NM_001324157.1 |
2461 |
Missense Mutation |
CAC,CGC |
H529R |
NP_001311086.1 |
NM_007130.3 |
2461 |
Missense Mutation |
CAC,CGC |
H565R |
NP_009061.1 |
NM_153380.3 |
2461 |
Missense Mutation |
CAC,CGC |
H565R |
NP_700359.1 |
XM_006724550.3 |
2461 |
Missense Mutation |
CAC,CGC |
H599R |
XP_006724613.1 |
XM_006724555.3 |
2461 |
Missense Mutation |
CAC,CGC |
H575R |
XP_006724618.1 |
XM_017029810.1 |
2461 |
Missense Mutation |
CAC,CGC |
H599R |
XP_016885299.1 |
XM_017029811.1 |
2461 |
Missense Mutation |
CAC,CGC |
H599R |
XP_016885300.1 |
XM_017029812.1 |
2461 |
Missense Mutation |
CAC,CGC |
H599R |
XP_016885301.1 |
XM_017029813.1 |
2461 |
Missense Mutation |
CAC,CGC |
H599R |
XP_016885302.1 |
XM_017029814.1 |
2461 |
Missense Mutation |
CAC,CGC |
H599R |
XP_016885303.1 |
XM_017029815.1 |
2461 |
Missense Mutation |
CAC,CGC |
H599R |
XP_016885304.1 |
XM_017029816.1 |
2461 |
Missense Mutation |
CAC,CGC |
H575R |
XP_016885305.1 |
XM_017029817.1 |
2461 |
Missense Mutation |
CAC,CGC |
H565R |
XP_016885306.1 |
View Full Product Details