Product Details

SNP ID
rs199534410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46500411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGGTGTGACTTTCCTCTGAAG[C/T]TTTTCCCACATTTACTACACTCATA
Phenotype
MIM: 300573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF674 PubMed Links

Gene Details

Gene
ZNF674
Gene Name
zinc finger protein 674
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039891.2 1576 Missense Mutation AAC,AGC N393S NP_001034980.1
NM_001146291.1 1576 Missense Mutation AAC,AGC N387S NP_001139763.1
NM_001190417.1 1576 Missense Mutation AAC,AGC N388S NP_001177346.1
XM_011543941.2 1576 Intron XP_011542243.1
XM_011543943.2 1576 Intron XP_011542245.1
XM_011543944.2 1576 Intron XP_011542246.1
XM_011543945.2 1576 Intron XP_011542247.1
XM_017029728.1 1576 Intron XP_016885217.1
XM_017029729.1 1576 Intron XP_016885218.1
XM_017029730.1 1576 Missense Mutation AAC,AGC N326S XP_016885219.1
XM_017029731.1 1576 Intron XP_016885220.1
XM_017029732.1 1576 Intron XP_016885221.1
XM_017029733.1 1576 Intron XP_016885222.1

View Full Product Details