Product Details

SNP ID
rs199797107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:108733001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCGAAGCGACAGCCGGGGCTGAG[G/T]ATGGGGAAAGGTCTCTCTCGAGGCT
Phenotype
MIM: 300904
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IRS4 PubMed Links

Gene Details

Gene
IRS4
Gene Name
insulin receptor substrate 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003604.2 3584 Missense Mutation TAC,TCC Y1115S NP_003595.1
XM_005262220.2 3584 Missense Mutation TAC,TCC Y1115S XP_005262277.1
XM_006724713.3 3584 Missense Mutation TAC,TCC Y1115S XP_006724776.1
XM_011531061.1 3584 Missense Mutation TAC,TCC Y1115S XP_011529363.1
Gene
LOC101928358
Gene Name
uncharacterized LOC101928358
There are no transcripts associated with this gene.

View Full Product Details