Product Details

SNP ID
rs199748306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:108087833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAGCATCTGGATTAGCCCCGCCT[G/T]CCAGTAACATGACAGCGATCTGGAA
Phenotype
MIM: 300663 MIM: 300880 MIM: 300620
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATG4A PubMed Links

Gene Details

Gene
ATG4A
Gene Name
autophagy related 4A cysteine peptidase
There are no transcripts associated with this gene.

Gene
PSMD10
Gene Name
proteasome 26S subunit, non-ATPase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002814.3 Intron NP_002805.1
NM_170750.2 Intron NP_736606.1
Gene
VSIG1
Gene Name
V-set and immunoglobulin domain containing 1
There are no transcripts associated with this gene.

View Full Product Details