Product Details

SNP ID
rs200352503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71616495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAGCAGCATCCACATCCGCTCC[C/T]GGAACTTGACCCCTACAAAGGCATA
Phenotype
MIM: 300369 MIM: 300574
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACRC PubMed Links

Gene Details

Gene
ACRC
Gene Name
acidic repeat containing
There are no transcripts associated with this gene.

Gene
CXCR3
Gene Name
C-X-C motif chemokine receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142797.1 706 Missense Mutation CAG,CGG Q373R NP_001136269.1
NM_001504.1 706 Missense Mutation CAG,CGG Q326R NP_001495.1
XM_005262256.3 706 Missense Mutation CAG,CGG Q336R XP_005262313.1
XM_005262257.3 706 Missense Mutation AGG,GGG R224G XP_005262314.1
XM_017029435.1 706 Missense Mutation CAG,CGG Q373R XP_016884924.1
XM_017029436.1 706 Missense Mutation AGG,GGG R214G XP_016884925.1

View Full Product Details