Product Details

SNP ID
rs199724832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:104013654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGAAGCCGGGGCACTTCGGTAC[A/G]CAGCATGGCTCCACGTCTCGGGCCA
Phenotype
MIM: 300507
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
H2BFWT PubMed Links

Gene Details

Gene
H2BFWT
Gene Name
H2B histone family member W, testis specific
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002916.4 34 Missense Mutation CGT,TGT R3C NP_001002916.3

View Full Product Details