Product Details
- SNP ID
-
rs201640492
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.Y:14622211 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCAATGTTCTTCTGTGGATAACTG[C/T]TCTTGCCATCAAGTTCACCCTCATT
- Phenotype
-
MIM: 400028
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NLGN4Y
PubMed Links
Gene Details
- Gene
- NLGN4Y
- Gene Name
- neuroligin 4, Y-linked
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001164238.1 |
321 |
Missense Mutation |
GCT,GTT |
A31V |
NP_001157710.1 |
NM_001206850.1 |
321 |
Intron |
|
|
NP_001193779.1 |
NM_014893.4 |
321 |
Missense Mutation |
GCT,GTT |
A31V |
NP_055708.3 |
XM_006724874.2 |
321 |
Missense Mutation |
GCT,GTT |
A31V |
XP_006724937.1 |
XM_011531424.2 |
321 |
Missense Mutation |
GCT,GTT |
A31V |
XP_011529726.1 |
XM_011531425.2 |
321 |
Missense Mutation |
GCT,GTT |
A31V |
XP_011529727.1 |
XM_011531426.2 |
321 |
Missense Mutation |
GCT,GTT |
A31V |
XP_011529728.1 |
XM_011531427.1 |
321 |
Missense Mutation |
GCT,GTT |
A31V |
XP_011529729.1 |
XM_011531428.1 |
321 |
Missense Mutation |
GCT,GTT |
A31V |
XP_011529730.1 |
XM_011531429.2 |
321 |
Missense Mutation |
GCT,GTT |
A31V |
XP_011529731.1 |
XM_011531430.2 |
321 |
Missense Mutation |
GCT,GTT |
A31V |
XP_011529732.1 |
XM_017030034.1 |
321 |
Missense Mutation |
GCT,GTT |
A31V |
XP_016885523.1 |
XM_017030035.1 |
321 |
Intron |
|
|
XP_016885524.1 |
XM_017030036.1 |
321 |
Missense Mutation |
GCT,GTT |
A31V |
XP_016885525.1 |
XM_017030037.1 |
321 |
Missense Mutation |
GCT,GTT |
A31V |
XP_016885526.1 |
XM_017030038.1 |
321 |
Missense Mutation |
GCT,GTT |
A31V |
XP_016885527.1 |
XM_017030039.1 |
321 |
Intron |
|
|
XP_016885528.1 |
XM_017030040.1 |
321 |
Intron |
|
|
XP_016885529.1 |
XM_017030041.1 |
321 |
Missense Mutation |
GCT,GTT |
A31V |
XP_016885530.1 |
View Full Product Details