Product Details

SNP ID
rs202214325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:52813167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGTGTGCCAGAGCCTCAACAA[A/G]AAGAACCACCAACTGAAAGTCAGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XAGE5 PubMed Links

Gene Details

Gene
XAGE5
Gene Name
X antigen family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130775.2 398 Missense Mutation AAA,GAA K34E NP_570131.1
XM_017029320.1 398 Missense Mutation AAA,GAA K34E XP_016884809.1

View Full Product Details