Product Details

SNP ID
rs202220241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103309819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGCCTAACGCGGAACCGCCTA[C/T]GGTTTCCTCTAGGCACACAGTATTC
Phenotype
MIM: 300691
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BEX2 PubMed Links

Gene Details

Gene
BEX2
Gene Name
brain expressed X-linked 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168399.1 354 Missense Mutation CAT,CGT H85R NP_001161871.1
NM_001168400.1 354 Missense Mutation CAT,CGT H84R NP_001161872.1
NM_001168401.1 354 Missense Mutation CAT,CGT H53R NP_001161873.1
NM_032621.3 354 Missense Mutation CAT,CGT H53R NP_116010.1

View Full Product Details