Product Details

SNP ID
rs202071965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:143026018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCTACAACCAACCATAATCATG[A/G]AAGAGCCAACTTCCAGCACCAACGA
Phenotype
MIM: 300667
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPANXN4 PubMed Links

Gene Details

Gene
SPANXN4
Gene Name
SPANX family member N4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009613.2 91 Intron NP_001009613.1
XM_017029543.1 91 Missense Mutation AAA,GAA K2E XP_016885032.1
XM_017029544.1 91 Missense Mutation AAA,GAA K2E XP_016885033.1

View Full Product Details