Product Details

SNP ID
rs202153801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119574940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAGCCGGAGTCAGCGGGGCTCC[G/T]TCCGAGAACAACATAATGGTGTGGA
Phenotype
MIM: 312180
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CXorf56 PubMed Links

Gene Details

Gene
CXorf56
Gene Name
chromosome X open reading frame 56
There are no transcripts associated with this gene.

Gene
UBE2A
Gene Name
ubiquitin conjugating enzyme E2 A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282161.1 329 UTR 5 NP_001269090.1
NM_003336.3 329 Silent Mutation CCG,CCT P28P NP_003327.2
NM_181762.2 329 Silent Mutation CCG,CCT P28P NP_861427.1

View Full Product Details