Product Details

SNP ID
rs201602104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:76175533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAAAGTAGATGACCAAATTTAC[G/T]CTGAGTTCCGGAAAAATTTTGAGAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PBDC1 PubMed Links

Gene Details

Gene
PBDC1
Gene Name
polysaccharide biosynthesis domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300888.1 420 Missense Mutation GCT,TCT A73S NP_001287817.1
NM_016500.3 420 Missense Mutation GCT,TCT A73S NP_057584.2

View Full Product Details