Product Details

SNP ID
rs200739971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70236108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGGTAAGGAACTGGTGTGTCT[A/G]GACCCACATCAGGGACTATTTCCCC
Phenotype
MIM: 300924
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AWAT1 PubMed Links

Gene Details

Gene
AWAT1
Gene Name
acyl-CoA wax alcohol acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013579.2 265 Nonsense Mutation TAG,TGG *75W NP_001013597.1
XM_017029295.1 265 Nonsense Mutation TAG,TGG *75W XP_016884784.1

View Full Product Details