Product Details

SNP ID
rs200794809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:105219112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTGATGAGGTTCGGAGTATCTC[C/T]TGTCTCTTGGGTGCTCCTGATGGTC
Phenotype
MIM: 300277 MIM: 300312
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL1RAPL2 PubMed Links

Gene Details

Gene
IL1RAPL2
Gene Name
interleukin 1 receptor accessory protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017416.1 Intron NP_059112.1
XM_011530905.2 Intron XP_011529207.1
XM_017029403.1 Intron XP_016884892.1
Gene
TEX13A
Gene Name
testis expressed 13A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291277.1 Intron NP_001278206.1
NM_031274.4 Intron NP_112564.1

View Full Product Details