Product Details
- SNP ID
-
rs200897550
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:124380740 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGCCTTCTTTGTTCCTTAGTCCA[A/G]GCCTGGGCCACTGCGCGCTGTCTGG
- Phenotype
-
MIM: 300490
MIM: 300588
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SH2D1A
PubMed Links
Gene Details
- Gene
- SH2D1A
- Gene Name
- SH2 domain containing 1A
There are no transcripts associated with this gene.
- Gene
- TENM1
- Gene Name
- teneurin transmembrane protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001163278.1 |
8799 |
Silent Mutation |
GCC,GCT |
A2665A |
NP_001156750.1 |
NM_001163279.1 |
8799 |
Silent Mutation |
GCC,GCT |
A2664A |
NP_001156751.1 |
NM_014253.3 |
8799 |
Silent Mutation |
GCC,GCT |
A2658A |
NP_055068.2 |
XM_011531230.2 |
8799 |
Silent Mutation |
GCC,GCT |
A2665A |
XP_011529532.1 |
XM_011531236.2 |
8799 |
Silent Mutation |
GCC,GCT |
A1900A |
XP_011529538.2 |
XM_011531237.2 |
8799 |
Silent Mutation |
GCC,GCT |
A1333A |
XP_011529539.1 |
XM_017029208.1 |
8799 |
Silent Mutation |
GCC,GCT |
A2698A |
XP_016884697.1 |
XM_017029209.1 |
8799 |
Silent Mutation |
GCC,GCT |
A2698A |
XP_016884698.1 |
XM_017029210.1 |
8799 |
Silent Mutation |
GCC,GCT |
A2698A |
XP_016884699.1 |
XM_017029211.1 |
8799 |
Silent Mutation |
GCC,GCT |
A2698A |
XP_016884700.1 |
XM_017029212.1 |
8799 |
Silent Mutation |
GCC,GCT |
A2698A |
XP_016884701.1 |
XM_017029213.1 |
8799 |
Silent Mutation |
GCC,GCT |
A2698A |
XP_016884702.1 |
XM_017029214.1 |
8799 |
Silent Mutation |
GCC,GCT |
A2658A |
XP_016884703.1 |
XM_017029215.1 |
8799 |
Intron |
|
|
XP_016884704.1 |
XM_017029216.1 |
8799 |
Intron |
|
|
XP_016884705.1 |
View Full Product Details