Product Details

SNP ID
rs201957890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103713708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACAGAACGATCAACCTTCTTGAA[C/G]TCCTTCCGTGGCCTACTGAGTGGAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GLRA4 PubMed Links

Gene Details

Gene
GLRA4
Gene Name
glycine receptor alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024452.2 408 Intron NP_001019623.2
NM_001172285.1 408 Intron NP_001165756.1
Gene
TMEM31
Gene Name
transmembrane protein 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182541.2 408 Missense Mutation CTC,GTC L73V NP_872347.2

View Full Product Details