Product Details

SNP ID
rs201216602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154432285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGAATGCCCTGGACCTGGCCC[C/G]CTCCTCACTGGTGCTTCCTGCCGTC
Phenotype
MIM: 300197 MIM: 300104
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP6AP1 PubMed Links

Gene Details

Gene
ATP6AP1
Gene Name
ATPase H+ transporting accessory protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001183.5 774 Missense Mutation CCC,CGC P128R NP_001174.2
XM_011531179.1 774 UTR 5 XP_011529481.1
Gene
CH17-340M24.3
Gene Name
uncharacterized protein BC009467
There are no transcripts associated with this gene.

Gene
GDI1
Gene Name
GDP dissociation inhibitor 1
There are no transcripts associated with this gene.

View Full Product Details