Product Details

SNP ID
rs200744728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:26217640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTATATGACCTTGTCAGCAAGCT[A/G]AAACTCCCCAACAATGGGAGGATTC
Phenotype
MIM: 300466
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGEB5 PubMed Links

Gene Details

Gene
MAGEB5
Gene Name
MAGE family member B5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271752.1 586 Silent Mutation CTA,CTG L113L NP_001258681.1

View Full Product Details