Product Details

SNP ID
rs201268079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103331598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAGATGAAGAAATGACAAGGGA[A/G]GGAGATGAGATGGAAAGGTGTTTGG
Phenotype
MIM: 300771
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCEAL7 PubMed Links

Gene Details

Gene
TCEAL7
Gene Name
transcription elongation factor A like 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152278.3 400 Silent Mutation GAA,GAG E65E NP_689491.1
XM_017029655.1 400 Silent Mutation GAA,GAG E65E XP_016885144.1

View Full Product Details