Product Details

SNP ID
rs201963962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154766249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTAATCTTGACAAGCCCTCTAA[C/T]CCCTCTTCCCATGAGGTGGTAGCCT
Phenotype
MIM: 300126
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DKC1 PubMed Links

Gene Details

Gene
DKC1
Gene Name
dyskerin pseudouridine synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142463.2 521 Silent Mutation AAC,AAT N99N NP_001135935.1
NM_001288747.1 521 Silent Mutation AAC,AAT N99N NP_001275676.1
NM_001363.4 521 Silent Mutation AAC,AAT N99N NP_001354.1
Gene
MIR664B
Gene Name
microRNA 664b
There are no transcripts associated with this gene.

Gene
SNORA36A
Gene Name
small nucleolar RNA, H/ACA box 36A
There are no transcripts associated with this gene.

Gene
SNORA56
Gene Name
small nucleolar RNA, H/ACA box 56
There are no transcripts associated with this gene.

View Full Product Details