Product Details
- SNP ID
-
rs200724711
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:63350551 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGCGCTAAAACTCTCTTATCGCGG[C/T]GCAGTTCTAGTCCATACACACTATC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SPIN4
PubMed Links
Gene Details
- Gene
- SPIN4
- Gene Name
- spindlin family member 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001012968.2 |
789 |
Missense Mutation |
CAC,CGC |
H90R |
NP_001012986.2 |
View Full Product Details