Product Details

SNP ID
rs201406258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30653581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTTTTGGGGCCATTGGTGGGGG[C/T]GGTGGACCAGGGCACCAGTTCGACG
Phenotype
MIM: 300474
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GK PubMed Links

Gene Details

Gene
GK
Gene Name
glycerol kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000167.5 223 Missense Mutation GCG,GTG A15V NP_000158.1
NM_001128127.2 223 Missense Mutation GCG,GTG A15V NP_001121599.1
NM_001205019.1 223 Missense Mutation GCG,GTG A15V NP_001191948.1
NM_203391.3 223 Missense Mutation GCG,GTG A15V NP_976325.1
XM_005274488.4 223 Intron XP_005274545.1
XM_006724483.2 223 Intron XP_006724546.1
XM_006724484.2 223 Intron XP_006724547.1
XM_006724485.2 223 Intron XP_006724548.1
XM_006724486.3 223 Intron XP_006724549.1
XM_011545491.2 223 Intron XP_011543793.1
XM_011545492.1 223 Intron XP_011543794.1
XM_011545493.2 223 Intron XP_011543795.1
XM_011545494.2 223 Intron XP_011543796.1
XM_017029409.1 223 Intron XP_016884898.1
XM_017029410.1 223 Intron XP_016884899.1
XM_017029411.1 223 Intron XP_016884900.1
XM_017029412.1 223 Intron XP_016884901.1

View Full Product Details