Product Details

SNP ID
rs201797267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:34639266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTACCTGCTGCAAAAAGCATGAC[C/T]GCAACAGGTCTATAGAAACGCCTTC
Phenotype
MIM: 300698
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM47 PubMed Links

Gene Details

Gene
TMEM47
Gene Name
transmembrane protein 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031442.3 607 Silent Mutation GCA,GCG A116A NP_113630.1

View Full Product Details