Product Details

SNP ID
rs200569560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:118974848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTCAGCAGCGACAACTTGGAGT[C/T]GGCGGAGCGAGGGGCATCAGCGGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LONRF3 PubMed Links

Gene Details

Gene
LONRF3
Gene Name
LON peptidase N-terminal domain and ring finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031855.2 235 Missense Mutation TCG,TTG S23L NP_001027026.1
NM_001289109.1 235 Intron NP_001276038.1
NM_024778.5 235 Missense Mutation TCG,TTG S23L NP_079054.3
XM_005262476.2 235 Missense Mutation TCG,TTG S23L XP_005262533.1
XM_011531398.2 235 Missense Mutation TCG,TTG S23L XP_011529700.1
XM_017029845.1 235 Missense Mutation TCG,TTG S23L XP_016885334.1

View Full Product Details