Product Details

SNP ID
rs200779428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:104186104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTGCCAGTGGGAGTGATGTACT[G/T]TCTGATGTCATACCCAGTATTCCAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM199X PubMed Links

Gene Details

Gene
FAM199X
Gene Name
family with sequence similarity 199, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207318.3 764 Silent Mutation CTG,CTT L152L NP_997201.1
XM_005262079.2 764 Silent Mutation CTG,CTT L152L XP_005262136.1
XM_017029270.1 764 UTR 5 XP_016884759.1

View Full Product Details