Product Details

SNP ID
rs201457607
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154404863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACATACGTCTCCATGTAGGTGCT[A/G]CGCCCCAGCTGGGGGCTGCTCAGGG
Phenotype
MIM: 300081 MIM: 312173 MIM: 300394
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DNASE1L1 PubMed Links

Gene Details

Gene
DNASE1L1
Gene Name
deoxyribonuclease I-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009932.2 907 Silent Mutation CGC,CGT R92R NP_001009932.1
NM_001009933.2 907 Silent Mutation CGC,CGT R92R NP_001009933.1
NM_001009934.2 907 Silent Mutation CGC,CGT R92R NP_001009934.1
NM_001303620.1 907 Silent Mutation CGC,CGT R92R NP_001290549.1
NM_006730.3 907 Silent Mutation CGC,CGT R92R NP_006721.1
XM_005277829.4 907 Silent Mutation CGC,CGT R92R XP_005277886.1
XM_011531122.2 907 Silent Mutation CGC,CGT R92R XP_011529424.1
XM_017029332.1 907 Silent Mutation CGC,CGT R92R XP_016884821.1
Gene
LOC107987332
Gene Name
uncharacterized LOC107987332
There are no transcripts associated with this gene.

Gene
RPL10
Gene Name
ribosomal protein L10
There are no transcripts associated with this gene.

Gene
SNORA70
Gene Name
small nucleolar RNA, H/ACA box 70
There are no transcripts associated with this gene.

Gene
TAZ
Gene Name
tafazzin
There are no transcripts associated with this gene.

View Full Product Details