Product Details
- SNP ID
-
rs201987772
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:117909301 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTATGCTTAATATAAATGCACTTAC[A/G]CAGTTCACCTGCTGCATTTCGCCCA
- Phenotype
-
MIM: 300655
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KLHL13
PubMed Links
Gene Details
- Gene
- KLHL13
- Gene Name
- kelch like family member 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001168299.1 |
2168 |
Missense Mutation |
CCC,TCC |
P459S |
NP_001161771.1 |
NM_001168300.1 |
2168 |
Missense Mutation |
CCC,TCC |
P450S |
NP_001161772.1 |
NM_001168301.1 |
2168 |
Missense Mutation |
CCC,TCC |
P440S |
NP_001161773.1 |
NM_001168302.1 |
2168 |
Missense Mutation |
CCC,TCC |
P440S |
NP_001161774.1 |
NM_001168303.1 |
2168 |
Missense Mutation |
CCC,TCC |
P414S |
NP_001161775.1 |
NM_033495.3 |
2168 |
Missense Mutation |
CCC,TCC |
P456S |
NP_277030.2 |
XM_011531409.2 |
2168 |
Missense Mutation |
CCC,TCC |
P462S |
XP_011529711.1 |
XM_011531410.2 |
2168 |
Missense Mutation |
CCC,TCC |
P462S |
XP_011529712.1 |
XM_011531411.1 |
2168 |
Missense Mutation |
CCC,TCC |
P456S |
XP_011529713.1 |
XM_017029950.1 |
2168 |
Missense Mutation |
CCC,TCC |
P440S |
XP_016885439.1 |
View Full Product Details