Product Details

SNP ID
rs200782951
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70926164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGATGCTCATTAGTGGGAGGAG[A/G]GGCAAAGCAGGCACCTGAAAACAAA
Phenotype
MIM: 300443
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SLC7A3 PubMed Links

Gene Details

Gene
SLC7A3
Gene Name
solute carrier family 7 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048164.2 1809 Silent Mutation CCC,CCT P545P NP_001041629.1
NM_032803.5 1809 Silent Mutation CCC,CCT P545P NP_116192.4
XM_017029912.1 1809 Silent Mutation CCC,CCT P545P XP_016885401.1

View Full Product Details