Product Details

SNP ID
rs201855156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119470446 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCACAGACTGTCACTGCTGTTGC[C/T]GGGTTGACTTCCTATCCATTTGACA
Phenotype
MIM: 300150
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A5 PubMed Links

Gene Details

Gene
SLC25A5
Gene Name
solute carrier family 25 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001152.4 788 Silent Mutation GCC,GCT A224A NP_001143.2
Gene
SLC25A5-AS1
Gene Name
SLC25A5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details