Product Details

SNP ID
rs201281829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153767122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGTAATCGACAGCCCTGACTGC[A/G]TGCCCTTCCGTGACCCAGCCGAGTG
Phenotype
MIM: 300214
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLXNB3 PubMed Links

Gene Details

Gene
PLXNB3
Gene Name
plexin B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163257.1 469 Missense Mutation ATG,GTG M122V NP_001156729.1
NM_005393.2 469 Missense Mutation ATG,GTG M99V NP_005384.2

View Full Product Details