Product Details

SNP ID
rs201098564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47233134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGATAGAGAGCCACAGCACGAG[A/G]AGCTGCCAGGCCTGGACAGCCAGTG
Phenotype
MIM: 311550 MIM: 300050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK16 PubMed Links

Gene Details

Gene
CDK16
Gene Name
cyclin dependent kinase 16
There are no transcripts associated with this gene.

Gene
USP11
Gene Name
ubiquitin specific peptidase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004651.3 220 Missense Mutation AAG,GAG K74E NP_004642.2
XM_005272674.3 220 Intron XP_005272731.1
XM_011543988.1 220 Intron XP_011542290.1

View Full Product Details