Product Details

SNP ID
rs200499663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:76177671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTCGGAACCGGGAAGGCTATAA[C/T]AAAGCTGTTTATATCAGTGTTCAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PBDC1 PubMed Links

Gene Details

Gene
PBDC1
Gene Name
polysaccharide biosynthesis domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300888.1 668 Missense Mutation ACA,ATA T118I NP_001287817.1
NM_016500.3 668 Silent Mutation AAC,AAT N155N NP_057584.2

View Full Product Details