Product Details

SNP ID
rs200552019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46500294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTTCTCTCCTGTATGAGTTCTA[C/T]GATGGACAGAGAGGTGTGACTTTCC
Phenotype
MIM: 300573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF674 PubMed Links

Gene Details

Gene
ZNF674
Gene Name
zinc finger protein 674
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039891.2 1693 Missense Mutation CAT,CGT H432R NP_001034980.1
NM_001146291.1 1693 Missense Mutation CAT,CGT H426R NP_001139763.1
NM_001190417.1 1693 Missense Mutation CAT,CGT H427R NP_001177346.1
XM_011543941.2 1693 Intron XP_011542243.1
XM_011543943.2 1693 Intron XP_011542245.1
XM_011543944.2 1693 Intron XP_011542246.1
XM_011543945.2 1693 Intron XP_011542247.1
XM_017029728.1 1693 Intron XP_016885217.1
XM_017029729.1 1693 Intron XP_016885218.1
XM_017029730.1 1693 Missense Mutation CAT,CGT H365R XP_016885219.1
XM_017029731.1 1693 Intron XP_016885220.1
XM_017029732.1 1693 Intron XP_016885221.1
XM_017029733.1 1693 Intron XP_016885222.1

View Full Product Details